ContactUsLinkComponent
DPD Chlorine Indicator Solution Ampules, ULR, Hach
DPD Chlorine Indicator Solution Ampules, ULR, Hach
Catalog # CA24932-20
Supplier:  Hach
CAS Number:  
undefined
DPD Chlorine Indicator Solution Ampules, ULR, Hach
Catalog # CA24932-20
Supplier:  Hach
Supplier Number:  2493220
CAS Number:  
Specifications
  • Chemical method:
    DPD (N,N-diethyl-p-phenylenediamine)
  • Description:
    DPD Chlorine Indicator Solution Ampules, ULR,
  • Method:
    8370 - DPD
  • Cat. no.:
    CA24932-20
  • Range:
    2 - 500 μg/L Cl₂
  • Supplier No.:
    24932-20
  • No. of Tests:
    20
Specifications
About this item
For ultra-low Total Chlorine Determination by the DPD method
Hach Method 8370. Range: 2 - 500 μg/L Cl2 Ampuled for stability.
Ordering information: Package of 20 1.5 mL ampules.
Hazardous Items:
Items with this mark may be considered hazardous under some shipping conditions.If necessary, we will change your selected shipping method to accomodate these items.