ContactUsLinkComponent
Dissolved Oxygen 3 Reagent Powder Pillows, Hach
Dissolved Oxygen 3 Reagent Powder Pillows, Hach
Catalog # CA987-99
Supplier:  Hach
CAS Number:  
undefined
Dissolved Oxygen 3 Reagent Powder Pillows, Hach
Catalog # CA987-99
Supplier:  Hach
Supplier Number:  98799
CAS Number:  
Specifications
  • Chemical method:
    Winkler
  • Description:
    Dissolved Oxygen 3 Reagent Powder Pillows
  • Size:
    60 mL Sample
  • Method:
    8161 - Winkler; 8209; 8211
  • Cat. no.:
    CA987-99
  • Range:
    Multiple ranges
  • Supplier No.:
    987-99
  • No. of Tests:
    100
Specifications
About this item
For determination of dissolved oxygen by the Winkler titration method.
Ordering information: Pack of 100 powder pillows.
Hazardous Items:
Items with this mark may be considered hazardous under some shipping conditions.If necessary, we will change your selected shipping method to accomodate these items.