Order Entry
Canada
ContactUsLinkComponent
 
m-FC Media, 2 ml Plastic Ampule, Hach
  CA97026-202
 :  Hach
 :  
undefined
m-FC Media, 2 ml Plastic Ampule, Hach
  CA97026-202
 :  Hach
 :  2373250
 :  

 

  • Description:
    m-FC media, Plastic Ampules
  • Size:
    2 mL
  • Cat. no.:
    CA97026-202
  • Supplier No.:
    2373250

 

 

m-FC 2mL plastic ampules

Save valuable time and money using this prepared media for membrane filtration methods.

Selectivity: Fecal Coliform Bacteria.

 : 50/pack.