Hardness 1 Buffer Solution, Hach
Fournisseur: Hach
Certificats
À propos de cet article
Buffer for hardness and chelant determination by titration, pH 10.1 ± 0.1 at 20°C
Hach methods 8213 and 8226. Also used in Hach Phenols method 8047.
Spécifications
- Method:EDTA Titration
Spécifications
Frequently Bought Together