ContactUsLinkComponent
Amino Acid F Reagent Solution, Hach
Amino Acid F Reagent Solution, Hach
Catalog # CA23864-42
Supplier:  Hach
CAS Number:  
undefined
Amino Acid F Reagent Solution, Hach
Catalog # CA23864-42
Supplier:  Hach
Supplier Number:  2386442
CAS Number:  
Specifications
  • Chemical method:
    Heteropoly blue
  • Description:
    Amino Acid F Reagent Solution
  • Size:
    100 mL
  • Method:
    8282 - Heteropoly Blue
  • Cat. no.:
    CA23864-42
  • Range:
    3 - 1000μg/L SiO₂
  • Supplier No.:
    23864-42
  • No. of Tests:
    100
Specifications
About this item
For determination of Ultra Low Range silica by the Heteropoly Blue method
Hach method 8282. Range: 3 - 1000μg/L SiO2. 100 mL bottle.