DPD Total Chlorine Reagent, SwifTest™ Dispenser Refill Vial, Hach
Catalog # CA21056-60
Supplier: Hach
CAS Number:
Specifications
- Chemical method:DPD (N,N-diethyl-p-phenylenediamine)
- Description:DPD Total Chlorine Reagent, SwifTest™ Dispenser Refill Vial
- Size:10 mL Sample
- Method:8167 - DPD
- Compliance:EPA - Yes
- Instrument:All except DR/3000, DR/2000
- Cat. no.:CA21056-60
- Range:0.02 - 2.00 mg/l Cl₂
- Supplier No.:21056-60
- No. of Tests:250
Specifications
About this item
For total chlorine determination by the DPD Total Chlorine method.
- Fast, easy reagent addition
- Just squeeze to add powder
For use with Hach SwifTest DPD dispenser. USEPA accepted for reporting for drinking and wastewater analysis using Hach Method 8167. Range: 0.02 - 2.00 mg/L Cl2.