Orders LinkContactUsLinkComponent
Ascorbic Acid Powder Pillows, Hach
Ascorbic Acid Powder Pillows, Hach
Ascorbic Acid Powder Pillows
Catalog # CA14577-99
Supplier:   Hach
Supplier #:  1457799
Ascorbic Acid Powder Pillows, Hach
Catalog # CA14577-99
Supplier:  Hach
CAS Number:  
Ascorbic Acid Powder Pillows, Hach
Ascorbic Acid Powder Pillows, Hach
Ascorbic Acid Powder Pillows
CAS Number:  
Price
$136.35
Log in for availability
Pack of 50
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
Reagent for aluminum determination by the aluminon method (Method 8012), manganese determination by the PAN method (Method 8149) and total chelant determination by titration (Method 8350).
Ordering information: Pack of 100 powder pillows.
Specifications
Specifications
  • Description:
    Ascorbic Acid Powder Pillows
Specifications
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/ca/en/login/register)