Chlorophosphonazo Indicator Solution for ULR Hardness, Hach
Catalog # CA25895-49
Supplier: Hach
CAS Number:
Specifications
- Chemical method:Chlorophosphonazo
- Description:Chlorophosphonazo Indicator Solution for ULR Hardness
- Size:500 mL bottle
- Method:8374 - Chlorophosphonazo
- Cat. no.:CA25895-49
- Range:4 - 1000 ug/L Ca & Mg as CaCO₃
- Supplier No.:25895-49
- No. of Tests:250
Specifications
About this item
For ultra low range hardness determination by the Chlorophosphonazo method.
- Convenience
- Performance
- Efficiency
Hach Method 8374. Range: 4 - 1000 ug/L Ca & Mg as CaCO3.
Ideal for use with Hach's Pour-Thru Cell. Requires CDTA Reagent, Cat. No. 97026-468.