Boss Head, Double Swivel
Catalog # 470137-842
Supplier: Wards
Specifications
- Description:Boss Head - Double Swivel
- Cat. no.:470137-842
- Supplier No.:CH0666
Specifications
About this item
Our swiveling boss head allows you to hold two rods at any angle.
Made of die cast zinc with the ability to hold rods up to 16 mm in diameter.