Boss Head, Square
470137-832
: Wards
- Description:Boss Head - Square
- Cat. no.:470137-832
- Supplier No.:CH0664
Perfect for Physics
This square boss head accommodates both square and tubular sections up to 16mm and holds them at right angles to each other. Holds a wide variety of objects including: rules, magnets, drawing boards, and more. The alloy body and plastic thumbscrews make it perfect for any physics lab.