Hardness (ULR) Reagent Set, Bulk Solution, Hach
Catalog # CA89231-140
Supplier: Hach
CAS Number:
Specifications
- Chemical method:Chlorophosphonazo
- Description:Hardness (ULR) Reagent Set, Bulk Solution
- Method:8374 - Chlorophosphonazo
- Instrument:All except DR/800, PC II
- Cat. no.:CA89231-140
- Range:4 - 1000 μg/l CaCO₃
- Supplier No.:2603101
- No. of Tests:250
Specifications
About this item
Reagent set ultra low range hardness determination by the Chlorophosphonazo method.
- Convenience
- Performance
- Efficiency
Hach Method 8374. Range: 4 - 1000µg/L Ca & Mg as CaCO3. Chlorophosphonazo reagent solution is packaged in a 500-mL bottle that is ideal for use with Hach's Pour-Thru Cell.