Distillation Apparatus, Vacuum Jacketed, Ace Glass
Supplier: Ace Glass
Certificates
About this item
This apparatus is a compact fractionation unit for distilling small quantities in a vacuum, and for ordinary separation
By use of the three-way stopcocks, a manifold arrangement is achieved, making it possible to remove receivers without disturbing the equilibrium of the system. Standard taper 10/30 thermometer joints are for use with 25mm immersion thermometers. Fabricated from borosilicate glass.
Column, including top joint, is vacuum jacketed. The indents are positioned to cause the vapor to follow a spiral path, adding to the efficiency of the unit. Approximately three theoretical plates at 60mL/hr. Hold up to 1. 2mL operating, 0.5mL static. Receiver capacity is 15mL.
Specifications
- Catalog No:
- 89055-624
- 89074-390
- Tubing size:
- 5/16 or 3/8" (C)
- 5/16 or 3/8" (C)