Still, 60 mm, Short Path, Ace Glass
Supplier: Ace Glass
Certificates
About this item
This short path still is designed to be effective with high boiling materials at low pressures, where moderate foaming, as well as slight bumping, can be tolerated
A fine tip adjustable gas inlet tube can be used for stirring, or as a foam breaker. A one-piece "cow" receiver with a 2.5mL measuring tube is supplied with complete unit. A 89074-356 "cow" receiver that accepts four 5 mL flasks or graduated receivers can be substituted for the one-piece "cow" receiver. Main joints are [ST] 14/20. The hose connections are for use with 5/16" I.D. tubing. Fabricated from borosilicate glass.
Ordering information: Complete apparatus includes 89074-364 still body; 89074-366 "cow" receiver with 3.0mL tubes; 89074-714 10mL distillation flask with [ST] 14/10 center neck and [ST] 10/18 side neck; 89074-350 gas inlet tube; and 89051-100 #7 Ace-Thred adapter with [ST] 10/10 joint. Optional components include 89074-356 "cow" receiver that accepts four 5mL flasks or graduated receivers; 5mL 89074-912 receiver flasks (four will be required); 5mL 89074-360 graduated receivers (0-1 mL x 0.1, 1-5mL x 0.2); and 89075-852 heating mantle (10mL). Individual components can be purchased separately.
Customers Who Bought This Also Bought