ContactUsLinkComponent
Moisture Resistance Indicator
Moisture Resistance Indicator
Catalog # 470137-772
undefined
Moisture Resistance Indicator
Catalog # 470137-772
Supplier Number:  GSK-944
Specifications
  • Description:
    Moisture Resistance Indicator
  • Cat. no.:
    470137-772
  • Supplier No.:
    GSK-944
Specifications
About this item
Check the Conductivity
Use this checker to measure dissolved solids in water. The greater the resistance, the less conductive the water; as resistance levels lessen, the materials that conduct electricity are elevated.The results are displayed via LED.