Distillation Apparatus, Markham Still, Ace Glass
Catalog #: 89055-622
Supplier: Ace Glass
Product available on GSA Advantage®
Small Business Enterprise
Specifications
- Description:Markham Still Distillation Apparatus
- Cat. no.:89055-622
- Supplier No.:6558-40
Specifications
About this item
This Markham Still distillation apparatus is used for volatile fatty acid tests
Fabricated from standard wall borosilicate glass tubing throughout. Hose connections on the still are for use with 3/8-inch I.D. tubing. Hose connections on the condenser are for use with 5/16-inch or 3/8-inch I.D. tubing.
Customers Who Bought This Also Bought