Orders LinkContactUsLinkComponent
Filter Support Assembly, Ace Glass
Filter Support Assembly, Ace Glass
Supplier:   Ace Glass
Filter Support Assembly, Ace Glass
Supplier:  Ace Glass
Certificates
Filter Support Assembly, Ace Glass
Filter Support Assembly, Ace Glass
12 Options Available Below
Product Details & Documents
These filter support assembly are used to collect dust particles in a stack sampling system.
  • Available in either 51 mm (2"), 76 mm (3"), or 102 mm (4") sizes
The filter assembly part A consists of [SJ] 28/15 outer joint bent perpendicular to flow path and part B consists of a seat for a fritted disc and a [SJ] 28/15 inner joint. Likewise, part C consists of a neoprene gasket used to seal filter paper and prevent leakage around the filter and part D with a Porosity B fritted disc.
Accessories information: Positive, two-piece aluminum clamps feature a gaskets and brass tightening bolts for securing the 76 mm (89059-262) or 102 mm (89059-264) size filter support assembly. The bolt head is flanged to secure the top half as the bolt is threaded into the bottom half.
Ordering information: Parts A, B, C, and D must be purchased separately. Two-piece clamps need to be ordered separately.
Product Family Options
Product Information
  • For use with
    Clamp size
    Availability
    Price
  • 89059-260
    Filter Support Assembly, Part A
    Filter Support Assembly, Part A
    Clamp 89060-266
    51 mm
    Log in for availability
    Log in for availability
    Each
    Each - $441.69
    $441.69
    New Customer Promotional Price
    Each
       $441.69
    Supplier Number: 7519-04
  • 89059-278
    Filter Support Assembly, Part B
    Filter Support Assembly, Part B
    Clamp 89060-266
    51 mm
    Log in for availability
    Log in for availability
    Each
    Each - $402.09
    $402.09
    New Customer Promotional Price
    Each
       $402.09
    Supplier Number: 7519-22
  • 89059-266
    Filter Support Assembly, Part C
    Filter Support Assembly, Part C
    Clamp 89060-266
    51 mm
    Log in for availability
    Log in for availability
    Each
    Each - $27.50
    $27.50
    New Customer Promotional Price
    Each
       $27.50
    Supplier Number: 7519-08
  • 89059-272
    Filter Support Assembly, Part D
    Filter Support Assembly, Part D
    Clamp 89060-266
    51 mm
    Log in for availability
    Log in for availability
    Each
    Each - $141.11
    $141.11
    New Customer Promotional Price
    Each
       $141.11
    Supplier Number: 7519-11
  • 89059-262
    Filter Support Assembly, Part A
    Filter Support Assembly, Part A
    Clamp 89055-042
    76 mm
    Log in for availability
    Log in for availability
    Each
    Each - $538.49
    $538.49
    New Customer Promotional Price
    Each
       $538.49
    Supplier Number: 7519-05
  • 89059-280
    Filter Support Assembly, Part B
    Filter Support Assembly, Part B
    Clamp 89055-042
    76 mm
    Log in for availability
    Log in for availability
    Each
    Each - $457.54
    $457.54
    New Customer Promotional Price
    Each
       $457.54
    Supplier Number: 7519-25
  • 89059-268
    Filter Support Assembly, Part C
    Filter Support Assembly, Part C
    Clamp 89055-042
    76 mm
    Log in for availability
    Log in for availability
    Each
    Each - $31.47
    $31.47
    New Customer Promotional Price
    Each
       $31.47
    Supplier Number: 7519-09
  • 89059-274
    Filter Support Assembly, Part D
    Filter Support Assembly, Part D
    Clamp 89055-042
    76 mm
    Log in for availability
    Log in for availability
    Each
    Each - $206.89
    $206.89
    New Customer Promotional Price
    Each
       $206.89
    Supplier Number: 7519-12
  • 89059-264
    Filter Support Assembly, Part A
    Filter Support Assembly, Part A
    Clamp 89055-044
    102 mm
    Log in for availability
    Log in for availability
    Each
    Each - $1,073.88
    $1,073.88
    New Customer Promotional Price
    Each
       $1,073.88
    Supplier Number: 7519-06
  • 89059-282
    Filter Support Assembly, Part B
    Filter Support Assembly, Part B
    Clamp 89055-044
    102 mm
    Log in for availability
    Log in for availability
    Each
    Each - $786.54
    $786.54
    New Customer Promotional Price
    Each
       $786.54
    Supplier Number: 7519-30
  • 89059-270
    Filter Support Assembly, Part C
    Filter Support Assembly, Part C
    Clamp 89055-044
    102 mm
    Log in for availability
    Log in for availability
    Each
    Each - $42.13
    $42.13
    New Customer Promotional Price
    Each
       $42.13
    Supplier Number: 7519-10
  • 89059-276
    Filter Support Assembly, Part D
    Filter Support Assembly, Part D
    Clamp 89055-044
    102 mm
    Log in for availability
    Log in for availability
    Each
    Each - $283.02
    $283.02
    New Customer Promotional Price
    Each
       $283.02
    Supplier Number: 7519-13
Accessories
Product Information
  • Catalog #
    For Use With
    Availability
    Price
  • Two Piece Clamp, Flat Flange, 160×120 mm
    Two Piece Clamp, Flat Flange, 160×120 mm
    89055-044
    Filter Support Assembly, 102 mm
    Log in for availability
    Each - $501.91
Product Information
  • Catalog #
    For Use With
    Availability
    Price
  • Two Piece Clamp, Flat Flange, 152×111 mm
    Two Piece Clamp, Flat Flange, 152×111 mm
    89055-042
    Filter Support Assembly, 76 mm
    Log in for availability
    Each - $532.54