Round-Bottom Flask with Beaded Tube Side Well, Ace Glass Incorporated
Supplier: Ace Glass
Certificates
About this item
This round-bottom flask has a center neck with [ST] outer joint and a deep beaded side well
The outer joint is tooled with reinforcement rings for added strength and stability. The I.D. of the side well is approximately 9 mm. Fabricated from borosilicate glass.
Ordering information: For a version of this flask with a side port rather than a deep side well, see 89056-522.
Frequently Bought Together