Corning® MCDB 131 1X, Corning
MCDB 131 1X
Supplier: Corning
Supplier #: 15-100-CV
Jump to:
- Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
- Return Policy
Product Details & Documents
MCDB 131 was originally developed by the researchers Knedler and Ham for the low-protein and serum free growth of human microvascular endothelial cells (HMVEC)
- Low endotoxin - < 0.25EU/mL
- Mycoplasma tested
- Formulated with cell culture grade WFI water
- Manufactured in a cGMP environment
Since its development, this medium has been used for this cell type and others such as liver, smooth muscle, cardiac myocytes. It is one of a group of MCDB media which were designed to provide a defined and optimally balanced nutritional environment for various mammalian cell types.
Documents
Certificates
Search by Lot NumberSpecifications
Specifications
- Description:MCDB 131 1X
- Cat. No.:CA45001-112
- Storage temperature:Refrigerate