JACKSON SAFETY® G60 Level 2 Cut Resistant Sleeves, KIMBERLY-CLARK PROFESSIONAL®
Catalog # CA89230-496
Supplier: Microflex
Specifications
- Type:G60 Level 2 cut resistant sleeves, with thumb hole
- Garment application:Laboratory
- Gender:Unisex
- Risk area:Cuts,Heat
- Color:Yellow
- Length:45.7 cm (18")
- Fabric:Kevlar®/cotton
- Cat. no.:CA89230-496
- Supplier No.:90070
Specifications
About this item
The new JACKSON SAFETY® G60 Level 2 Cut Resistant Sleeves are a great choice for working environments that pose a risk for minor cuts, abrasions, and burns
Kevlar® outer layer and cotton inner layer. ANSI Level 2/EN 388 Level 2 cut resistance. Comfortable fit with reliable breathability. Suitable for intermittent heat contact up to 212° F (sleeve must not come into contact with a naked flame). Meets EN 407: Thermal Hazards. 18" length. Yellow. Typical uses include metal fabrication, glass handling, automotive assembly, stamping, and assembly.