Order Entry
Canada
ContactUsLinkComponent
Clamp Holder, 13 mm Grip, Ace Glass
Clamp Holder, 13 mm Grip, Ace Glass
  71001-712
 :  Ace Glass
Product available on GSA Advantage®
Small Business Enterprise
undefined
Clamp Holder, 13 mm Grip, Ace Glass
  71001-712
 :  Ace Glass
 :  11086-01
Product available on GSA Advantage®
Small Business Enterprise

 

  • Description:
    Stainless Steel Clamp Holder
  • Cat. no.:
    71001-712
  • Supplier No.:
    11086-01

 

 

This stainless steel lab frame hook connector allow one-handed assembly of two components with one adjustment screw

It has a 13mm grip.