Order Entry
Canada
ContactUsLinkComponent
Potassium hydroxide 50% (w/v) in aqueous solution, VWR Chemicals BDH®
Corrosive
Potassium hydroxide 50% (w/v) in aqueous solution, VWR Chemicals BDH®
Catalog # BDH7622-1
Supplier:  VWR International
CAS Number:  
undefined
Corrosive
Potassium hydroxide 50% (w/v) in aqueous solution, VWR Chemicals BDH®
Catalog # BDH7622-1
Supplier:  VWR International
CAS Number:  

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/ca/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Specifications

  • Pack type:
    Poly bottle
  • Size:
    1 L
  • Cat. no.:
    BDH7622-1
  • Supplier No.:
    BDHVBDH7622-1

Specifications

Specification Test Results

  • Appearance/color:
    Clear liquid
  • Shelf-life:
    2 years
  • Assay (w/v):
    48.00 - 52.00 %

Specification Test Results

About this item

 :
 :56.11 g/mol
Storage Temperature:Ambient
EINECS:215-181-3
UN:1814
ADR:8,II
 :GHS05
CAS Number:1310-58-3
Formula:KOH
MDL Number:MFCD00003553