Order Entry
Canada
Orders LinkContactUsLinkComponent
Phosphate, 0.5 M Buffer Solution, pH 7.2, Thermo Scientific

Phosphate, 0.5 M Buffer Solution, pH 7.2, Thermo Scientific

Phosphate, 0.5 M Buffer Solution, pH 7.2, Thermo Scientific
Certificates
Phosphate, 0.5 M Buffer Solution, pH 7.2, Thermo Scientific

2 Options Available Below

Product Details & Documents

Liquid

CAS No.: 7558-80-7
Physical Form: Liquid
MDL No.: MFCD00003527

Documents

Specifications for Product Family

Specifications

Specifications

  • Catalog No:
  • CAAAJ63974-AK
  • CAAAJ63974-AP
  • MDL No.:
  • MFCD00003527
  • MFCD00003527

Specifications

Product Family Options

Product Information

  • Size
    Availability
    Price
  • 250 mL
    Log in for availability
    Log in for availability
    Each (250ml)
    Each (250ml) - $201.11
    $201.11
    Each (250ml)
       $201.11
    Minimum order quantity is 1

    Supplier Number: J63974-AK
    Specifications:
    More
    Specifications
    Cat. No.
    CAAAJ63974-AK
    MDL No.
    MFCD00003527
  • 500 mL
    Log in for availability
    Log in for availability
    Each (500ml)
    Each (500ml) - $276.48
    $276.48
    Each (500ml)
       $276.48
    Minimum order quantity is 1

    Supplier Number: J63974-AP
    Specifications:
    More
    Specifications
    Cat. No.
    CAAAJ63974-AP
    MDL No.
    MFCD00003527
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.