Order Entry
Canada
Orders LinkContactUsLinkComponent
Pectinase
Pectinase
Certificates

2 Options Available Below

Product Details & Documents

Powder

CAS No.: 9032-75-1 Physical Form: Powder Appearance: Off-white to pale brown MDL No.: MFCD00131809

Specifications for Product Family

Specifications

Specifications

  • Catalog No:
  • CAAAJ63408-MD
  • CAAAJ63408-03
  • Enzyme name:
  • Pectinase
  • Pectinase
  • CAS No.:
  • 9032-75-1
  • 9032-75-1
  • Form:
  • Powder
  • Powder

Specifications

Product Family Options

Product Information

  • Size
    Availability
    Price
  • 250 mg
    Log in for availability
    Log in for availability
    Each (250mg)
    Each (250mg) - $319.98
    $319.98
    Each (250mg)
       $319.98
    Minimum order quantity is 1

    Supplier Number: J63408-MD
    Specifications:
    More
    Specifications
    Cat. No.
    CAAAJ63408-MD
    Enzyme name
    Pectinase
    CAS No.
    9032-75-1
    Form
    Powder
  • 1 g
    Log in for availability
    Log in for availability
    Each (1g)
    Each (1g) - $829.94
    $829.94
    Each (1g)
       $829.94
    Minimum order quantity is 1

    Supplier Number: J63408-03
    Specifications:
    More
    Specifications
    Cat. No.
    CAAAJ63408-03
    Enzyme name
    Pectinase
    CAS No.
    9032-75-1
    Form
    Powder
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.