Transformation buffer-2, pH 7.5, Thermo Scientific
Supplier: Thermo Scientific Chemicals
Certificates
About this item
Liquid
Storage Temperature: 2°C to 8°C
Physical Form: Liquid
For transformation of competent cells
Related Information
Frequently Bought Together