TAPS, 0.2 M Buffer Solution, pH 8.5, Thermo Scientific
Catalog # CAAAJ63268-AE
Supplier: Thermo Scientific Chemicals
CAS Number:
Specifications
- Size:100 mL
- MDL No.:MFCD00069903
- Cat. no.:CAAAJ63268-AE
- Supplier No.:J63268-AE
Specifications
About this item
Liquid
CAS No.: 91000-53-2
Physical Form: Liquid
MDL No.: MFCD00069903