Thrombin Buffer, pH 8.0, Thermo Scientific
Supplier: Thermo Scientific Chemicals
Jump to:
About this item
Liquid
Storage Temperature: 2°C to 8°C
Physical Form: Liquid
Appearance: Colorless
Odor: Odorless
2 Options Available Below
Product Details & Documents
Liquid
Storage Temperature: 2°C to 8°C
Physical Form: Liquid
Appearance: Colorless
Odor: Odorless



