Order Entry
Canada
ContactUsLinkComponent
Arthrobacter luteus Yeast lytic enzyme
Arthrobacter luteus Yeast lytic enzyme
Certificates
Arthrobacter luteus Yeast lytic enzyme
Certificates

Specifications

  • CAS No.:
    37340-57-1
  • Species:
    Arthrobacter luteus
  • Storage conditions:
    2...8 °C
  • Form:
    Off-White lyophilized powder
  • MDL No.:
    MFCD00131560
  • Enzyme name:
    Yeast lytic enzyme

Specifications

Related Information

Frequently Bought Together