TRIS-buffered saline (TBS, 10X), with 1% Tween 20, Thermo Scientific
Supplier: Thermo Scientific Chemicals
Certificates
About this item
Liquid
Physical Form: Liquid
MDL No.: MFCD00678198
Specifications
- MDL No.:MFCD00678198
Specifications
Related Information
Frequently Bought Together