Phosphate, 0.2 M Buffer Solution, pH 7.2, Thermo Scientific
Supplier: Thermo Scientific Chemicals
Certificates
About this item
Liquid
CAS No.: 7558-80-7
Physical Form: Liquid
MDL No.: MFCD00003527
Specifications
- MDL No.:MFCD00003527
Specifications
Related Information
Frequently Bought Together