Order Entry
Canada
Orders LinkContactUsLinkComponent
 
GTE buffer
 
GTE buffer

 

 

Storage temperature: 2 to 8°C

  • Suitable for miniprep and DNA purification

Contains 25 mM Tris, 50 mM glucose, and 10 mM EDTA.

Product Family Options

 

  • Size
     
     
  • 100 mL
       
       
    Each (100ml)
    Each (100ml) - $54.80
    $54.80
    Each (100ml)
       $54.80
     

     : J62597-AE
  • 250 mL
       
       
    Each (250ml)
    Each (250ml) - $102.88
    $102.88
    Each (250ml)
       $102.88
     

     : J62597-AK