Order Entry
Canada
Orders LinkContactUsLinkComponent
 

HEPPSO, 0.2 M Buffer Solution, pH 8.0, Thermo Scientific

HEPPSO, 0.2 M Buffer Solution, pH 8.0, Thermo Scientific
Certificates
HEPPSO, 0.2 M Buffer Solution, pH 8.0, Thermo Scientific

1 Options Available Below

Product Details & Documents

Liquid

CAS No.: 68399-78-0
Physical Form: Liquid
MDL No.: MFCD00038351

Specifications for Product Family

Specifications

Specifications

  • Catalog No:
  • CAAAJ62529-AE
  • CAAAJ62529-AK
  • MDL No.:
  • MFCD00038351
  • MFCD00038351

Specifications

Product Family Options

Product Information

  • Size
    Availability
    Price
  • 100 mL
    Discontinued
    Supplier Number: -
    Specifications:
    More
    Specifications
    Cat. No.
    CAAAJ62529-AE
    MDL No.
    MFCD00038351
  • 250 mL
    Log in for availability
    Log in for availability
    Each (250ml)
    Each (250ml) - $221.16
    $221.16
    Each (250ml)
       $221.16
     

    Supplier Number: J62529-AK
    Specifications:
    More
    Specifications
    Cat. No.
    CAAAJ62529-AK
    MDL No.
    MFCD00038351