Schiff reagent
500 ml
Supplier: Thermo Scientific Chemicals
: J62171-AP
Jump to:
Product Details & Documents
Used for detection of glycoproteins in polyacrylamide gels
Intended for use in Sigma Periodic Acid Staining Procedure, 395. Kit is intended for use with blood, bone marrow, tissue touch preparations or routine tissue sections. It is used for staining glycoproteins in polyacrylamide gels. Schiff's reagent is also sometimes used to dye organic tissues. It is been widely used for detection of a certain functional group in chemical solutions.
Used for detection of glycoproteins in polyacrylamide gels
: Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Keep away from strong oxidizing agents, strong alkalis and acids.
- MDL Number:MFCD00081985
- Density:1.1 g/cm³ (20 °C)
- Storage Temperature:Ambient
Specifications
Specifications
- Size:500 ml



