Order Entry
Canada
Orders LinkContactUsLinkComponent
LDS-sample buffer (4X), non-reducing, Thermo Scientific

LDS-sample buffer (4X), non-reducing, Thermo Scientific

LDS-sample buffer (4X), non-reducing, Thermo Scientific
Certificates
LDS-sample buffer (4X), non-reducing, Thermo Scientific

2 Options Available Below

Product Details & Documents

LDS-sample buffer (4X), non-reducing is used in SDS-polyacrylamide gel electrophoresis (SDS-PAGE). It is used in denaturing gels and is compatible with both coomassie and silver staining and Western blotting applications.

Physical Form: Liquid
Appearance: Blue

This buffer contains: 988 mM Tris-HCl, 2.04 mM EDTA, 8% LDS, 40% glycerol, 0.88% Coomassie blue and 0.7 mM phenol red at pH 8.5.

Product Family Options

Product Information

  • Size
    Availability
    Price
  • 25 mL
    Log in for availability
    Log in for availability
    Each (25ml)
    Each (25ml) - $155.32
    $155.32
    Each (25ml)
       $155.32
    Minimum order quantity is 1

    Supplier Number: J61894-AC
  • 50 mL
    Log in for availability
    Log in for availability
    Each (50ml)
    Each (50ml) - $258.86
    $258.86
    Each (50ml)
       $258.86
    Minimum order quantity is 1

    Supplier Number: J61894-AD