Sodium Phosphate, 0.5 M Buffer Solution, pH 7.5, Thermo Scientific
Supplier: Thermo Scientific Chemicals
Certificates
About this item
Liquid
Physical Form: Liquid
MDL No.: MFCD00146206
Specifications
- MDL No.:MFCD00146206
Specifications
Related Information
Frequently Bought Together