MOBS, 0.2 M Buffer Solution, pH 7.0, Thermo Scientific
Liquid
CAS No.: 115724-21-5
Storage Temperature: 2°C to 8°C
Physical Form: Liquid
Appearance: Colorless
MDL No.: MFCD00674916
- MDL No.:MFCD00674916
Frequently Bought Together
Liquid
CAS No.: 115724-21-5
Storage Temperature: 2°C to 8°C
Physical Form: Liquid
Appearance: Colorless
MDL No.: MFCD00674916