Phosphate-Buffered Saline (PBS, 1X), Sterile Filtered, Thermo Scientific
Catalog # CAAAJ61196-AP
Supplier: Thermo Scientific Chemicals
CAS Number:
Specifications
- Size:500 mL
- Cat. no.:CAAAJ61196-AP
- Supplier No.:J61196-AP
Specifications
About this item
Liquid
Physical Form: Liquid