Carbonate, 0.5 M Buffer Solutions, Thermo Scientific
Supplier: Thermo Scientific Chemicals
Certificates
About this item
Liquid
CAS No.: 497-19-8
Odor: Odorless
MDL No.: MFCD00003494
Specifications
- MDL No.:MFCD00003494
Specifications
Related Information
Frequently Bought Together