Sucrose, 80%, in MNE buffer, MES + NaCl + EDTA, Thermo Scientific
Supplier: Thermo Scientific Chemicals
Certificates
About this item
Liquid
Physical Form: Viscous liquid
MDL No.: MFCD00006626
Specifications
- MDL No.:MFCD00006626
Specifications
Related Information
Frequently Bought Together