Aphidicolin
Supplier: Thermo Scientific Chemicals
Jump to:
About this item
Cyrstalline powder
A DNA polymerase inhibitor. Blocks the cell cycle at the early S-phase
A DNA polymerase inhibitor. Blocks the cell cycle at the early S-phase
Synonyms: (3R,4R,4aR,6aS,8R,9R,11aS,11bS)-4,9-Bis(hydroxymethyl)-4,11b-dimethyltetradecahydro-8,11a-methanocyclohepta[a]naphthalene-3,9-diol
- CAS Number:38966-21-1
- Formula:C₂₀H₃₄O₄
- MDL Number:MFCD00083214
- Molecular Weight:338.488 g/mol
- Merck Index:14,727
- Melting Pt:224 °C
- Storage Temperature:Refrigerator
2 Options Available Below
Product Details & Documents
Cyrstalline powder
A DNA polymerase inhibitor. Blocks the cell cycle at the early S-phase
A DNA polymerase inhibitor. Blocks the cell cycle at the early S-phase
Synonyms: (3R,4R,4aR,6aS,8R,9R,11aS,11bS)-4,9-Bis(hydroxymethyl)-4,11b-dimethyltetradecahydro-8,11a-methanocyclohepta[a]naphthalene-3,9-diol
- CAS Number:38966-21-1
- Formula:C₂₀H₃₄O₄
- MDL Number:MFCD00083214
- Molecular Weight:338.488 g/mol
- Merck Index:14,727
- Melting Pt:224 °C
- Storage Temperature:Refrigerator
Documents
Product Family Options
Product Information
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



