Denaturing lysis buffer, Thermo Scientific
Supplier: Thermo Scientific Chemicals
Jump to:
About this item
Liquid
Physical Form: Liquid
Appearance: Clear liquid
pH: 7.4
1 Options Available Below
Product Details & Documents
Liquid
Physical Form: Liquid
Appearance: Clear liquid
pH: 7.4
Documents
Product Family Options
Product Information
- SizeAvailabilityPrice
- Supplier Number: J60132-AK
- 500 mLDiscontinued-Supplier Number: -
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



