SP Bel-Art Poxygrid® Centrifuge Tube Rack, 500 ml, Bel-Art Products, a part of SP
89093-450
- Description:Poxygrid® 500mL Centrifuge Tube Rack
- Dimensions:20.3L x 20.3W x 7.6H cm (8 x 8 x 3")
- Cat. no.:89093-450
- Supplier No.:198560500
The only wire rack that holds 500 mL conical centrifuge tubes.
- Epoxy-coated steel wire construction
- Conical-bottomed centrifuge tubes
- Autoclavable
Support, transport, and store up to four 500 mL conical bottom centrifuge tubes. Epoxy-coated steel wire. Steam autoclavable at 121ºC (250ºF).