Small pH Meter Supplies Organizer, PVC, TrippNT
Catalog #: 11376-298
Supplier: TrippNT
Minority or Woman-Owned Business Enterprise
Small Business Enterprise
Specifications
- Depth:6"
- Width:12"
- Description:Two-Shelf Organizer with Safety Lip
- Height:12"
- Weight:1 kg (2 lbs.)
- Supplier No.:50006
Specifications
About this item
White PVC small pH meter supplies organizer is a great way to store pH meter supplies.
- Made of chemically resistant PVC
- The open design and 6" deep shelves make it easy to get larger items in and out
Keep buffers, probes, sample cups, squeeze bottles, waste beakers and wipers all together in one organizer. This workstation is compact, its two shelf design creates high storage capacity with a small footprint. Built-in wiper dispenser and six hole tool holder. The shelves have a 1" safety lip, keeping supplies securely in their place, and each unit has multiple compartments to store supplies.
Customers Who Bought This Also Bought