Storage Bin with Eight Adjustable Compartments and Two Shelves, PVC, TrippNT
Catalog # 11376-240
Supplier: TrippNT
Minority or Woman-Owned Business Enterprise
Small Business Enterprise
Specifications
- Color:White
- Description:Storage Bin
- Cat. no.:11376-240
- W×D×H:30 x 25 x 51 cm (12 x 10 x 20")
- Supplier No.:50152
Specifications
About this item
White PVC storage bin with 8 adjustable compartments and 2 shelves creates storage to fit supply volume.
- Sorts, stores, organize and protects supplies
- With safety lips
This storage bin is constructed of durable white PVC to stand up to demanding work environments. The 8 adjustable bins and shelves store gloves, wipers, reagents, vials, towels, glassware and other supplies.