Distillation Column, Jacketed with Indents, Ace Glass Incorporated
89074-410
Product available on GSA Advantage®
Small Business Enterprise
- Description:Distillation Column
- Cat. no.:89074-410
- Supplier No.:9342-01
This indented distillation column has a jacket with a length of 250mm
It has a [ST] 14/20 outer joint at top and a [ST] 14/20 inner joint at bottom. There are hose connections on each side of the column. The indents can support glass beads or other packing. The hose connections are for use with 5/16-inch or 3/8-inch I.D. tubing. Fabricated from borosilicate glass.