Graduated Cylinders, Octagonal Base, Ace Glass Incorporated
Supplier: Ace Glass
Certificates
About this item
These polypropylene cylinders comes with an octagonal base
Ideal for rugged industrial and school lab use. With single scale graduations for easier, and quicker reading. No meniscus means easy-to-read liquid level. Large octagonal base provides good stability. 10mL size has conical top for easy filling. Autoclavable, however repeated autoclaving may affect accuracy. May also be sterilized by standard chemical methods.
Specifications
- Catalog No:
- 71000-248
- 71000-236
- 71000-244
- 71000-242
- 71000-240
- 71000-238
- 71000-246
- Form:
- Tall
- Tall
- Tall
- Tall
- Tall
- Tall
- Tall