To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
- Antibody type:Primary
- Antigen name:WNK lysine deficient Protein kinase 4
- Antigen symbol:WNK4
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human
- Western blot:Yes
- Size:100 μl
- Antigen synonyms:PHA2B|PRKWNK4
- Gene ID:65266
- Concentration:0.92 mg/ml
- Immunogen:A synthetic peptide sequence made to a region of the human protein between residues 200-300.
- Purification:Antigen affinity purification
- Storage temperature:Keep as concentrated solution. Store at 4 °C short term. For extended storage aliquot and store at –20 °C or below. Avoid freeze-thaw cycles.
- Tested applications:WB
- Cat. no.:89362-938
- Supplier No.:GTX30604
Specifications
About this item
Rabbit polyclonal antibody to WNK4
For WB: Use at a concentration of 2 ug/ml (ECL). This antibody is useful for Western blot where a band is seen at ~135-155 kD. Not tested in other applications. Optimal dilutions/concentrtions should be determined by the researcher.
Type: Primary
Antigen: WNK4
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human