Order Entry
ContactUsLinkComponent
Anti-TRP1 Mouse Monoclonal Antibody [clone: 401]
Anti-TRP1 Mouse Monoclonal Antibody [clone: 401]
Catalog # 89363-222
Supplier:  Genetex
Anti-TRP1 Mouse Monoclonal Antibody [clone: 401]
Catalog # 89363-222
Supplier:  Genetex
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:

  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address

* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    Trypsin
  • Antigen symbol:
    TRP1
  • Clonality:
    Monoclonal
  • Clone:
    401
  • Conjugation:
    Unconjugated
  • ELISA:
    Yes
  • Host:
    Mouse
  • Isotype:
    IgG2b
  • Reactivity:
    Human
  • Size:
    100 µg
  • Antigen synonyms:
    Protease Serine 1 Preproprotein|Nonfunctional Trypsin 1|Protease Serine 1|Cationic Trypsinogen|Serine Protease 1|Digestive Zymogen|T|TRY1|TRP1
  • Immunogen:
    Full length native protein (purified) (Human pancreatic).
  • Purification:
    purified by Protein G chromatography
  • Storage buffer:
    PBS, pH 7.4, containing no preservatives
  • Storage temperature:
    –20 °C
  • Tested applications:
    ELISA
  • Cat. no.:
    89363-222
  • Supplier No.:
    GTX40555

Specifications

About this item

Trypsin is a 24 kDa enzyme of the serine proteinase family. It is produced in the pancreas as an inactive precursor, trypsinogen, but the active enzyme is located in the gastrointestinal tract where it degrades proteins to large peptides. Trypsin prefers lysine and arginine residues. One of the substrates for trypsin is chymotrypsinogen which is cleaved to produce active chymotrypsin. High levels of immunoreactive trypsin in the bloodstream can indicate pancreatic malfunction and this indicator is used to screen for and diagnose Cystic Fibrosis.

Type: Primary
Antigen: TRP1
Clonality: Monoclonal
Clone: 401
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human

Customers Who Bought This Also Bought