Order Entry
ContactUsLinkComponent
Anti-TLR2 Mouse Monoclonal Antibody [clone: 11G7]
Catalog #: 89363-524
Supplier:  Genetex
Price
$1,036.72
Log in for availability
Unit of Measure Each
Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:

  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address

* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    Toll-like Receptor 2
  • Clonality:
    Monoclonal
  • Clone:
    11G7
  • Gene ID:
    7097
  • Host:
    Mouse
  • Isotype:
    IgG1
  • Reactivity:
    Human
  • Antigen symbol:
    TLR2
  • Conjugation:
    PE (Phycoerythrin)
  • Flow cytometry:
    Yes
  • Antigen synonyms:
    CD282
    TIL4
    Toll/interleukin-1 receptor-like protein 4
  • Storage buffer:
    Phosphate buffered saline containing 0.09% Sodium Azide (NaN3) 1% Bovine Serum Albumin 5% Sucrose
  • Storage temperature:
    Store at +4°C. DO NOT FREEZE. This product should be stored undiluted. This product is photosensitive and should be protected from light. Should this product contain a precipitate we recommend microcentrifugation before use.
  • Tested applications:
    Fluorescence-Activated Cell Sorting
  • Purification:
    Purified IgG prepared by affinity chromatography on Protein G from tissue culture supernatant
  • Cat. No.:
    89363-524
  • Size:
    100 test

Specifications

About this item