Order Entry
ContactUsLinkComponent
Anti-Sperm Fertilization 56 Mouse Monoclonal Antibody
Anti-Sperm Fertilization 56 Mouse Monoclonal Antibody
Catalog #: 89360-296
Supplier:  Genetex
Anti-Sperm Fertilization 56 Mouse Monoclonal Antibody
Catalog #: 89360-296
Supplier:  Genetex
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:

  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address

* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    Sperm Fertilization 56
  • Clonality:
    Monoclonal
  • Host:
    Mouse
  • Isotype:
    IgG3
  • Reactivity:
    Mouse
  • Antigen symbol:
    Sperm Fertilization 56
  • Conjugation:
    Unconjugated
  • ELISA:
    Yes
  • ImmunoChemistry:
    Yes
  • Western blot:
    Yes
  • Storage buffer:
    Preservative: None. Constituents: PBS. pH 7.2.
  • Storage temperature:
    Keep as concentrated solution. Store at 4 °C short term. For extended storage aliquot and store at –20 °C or below. Avoid freeze-thaw cycles.
  • Immunogen:
    Mixture of native and formaldehyde zinc fixed sperm purified through a Percoll gradient.
  • Tested applications:
    ELISA, IHC, WB
  • Purification:
    Protein G purified
  • Cat. no.:
    89360-296
  • Supplier No.:
    GTX17359

Specifications

About this item

Binds to ZP3 glycoprotein in egg zona pellucida. Believed to be involved in interactions between the sperm acrosome and egg zona pellucida during and immediately following the acrosome reaction.

Type: Primary
Antigen: Sperm Fertilization 56
Clonality: Monoclonal
Clone:
Conjugation:
Epitope:
Host: Mouse
Isotype: IgG3
Reactivity: Mouse

Customers Who Bought This Also Bought