Anti-SDCCAG3 Rabbit Polyclonal Antibody
Catalog # 89354-526
Supplier: Genetex
Specifications
- Antibody type:Primary
- Antigen name:Serologically Defined Colon Cancer Antigen 3
- Antigen symbol:SDCCAG3
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human
- Western blot:Yes
- Size:100 μl
- Gene ID:10807
- Antigen synonyms:NY-CO-3
- Storage buffer:0.1M Tris, 0.1M Glycine, 10% Glycerol (pH7). 0.01% Thimerosal was added as a preservative.
- Molecular weight:48 kDa
- Storage temperature:Keep as concentrated solution. Aliquot and store at –20 °C or below. Avoid multiple freeze-thaw cycles.
- Concentration:1mg/ml
- Immunogen:Recombinant protein encompassing a sequence within the center region of human SDCCAG3. The exact sequence is proprietary.
- Tested applications:WB
- Purification:Purified by antigen-affinity chromatography.
- Cat. no.:89354-526
- Supplier No.:GTX106180
Specifications
About this item
Rabbit Polyclonal antibody to SDCCAG3 (serologically defined colon cancer antigen 3)
Suggested starting dilutions are as follows: WB: 1:500-1:3000. Not yet tested in other applications. Optimal working dilutions should be determined experimentally by the end user.
Type: Primary
Antigen: SDCCAG3
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human